Learn More
Electron Microscopy Sciences Dri-Bath™ Solid Aluminum Block, Wells
SDP
Catalog No. 5032252
Encompass_Preferred
These versatile soild aluminum blocks have an anondized coating that resists the harshest environments. Provide maximum heat retention. All blocks include thermometer well for easily measuring block temperature. Fit all our Modular Block Dri-Baths.
Catalog No. 50-322-52 Supplier Electron Microscopy Sciences Supplier No. 6312430
These versatile soild aluminum blocks have an anondized coating that resists the harshest environments. Provide maximum heat retention. All blocks include thermometer well for easily measuring block temperature. Fit all our Modular Block Dri-Baths.