Learn More
Cole-Parmer Cole-Parmer Solid Black Rubber Stoppers, Standard Size 13.5; 3/Pk
Catalog No. NC1489330 Shop All Cole-Parmer Products
Encompass_Preferred
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.
Catalog No. NC1489330 Supplier Cole-Parmer Supplier No. 6299046
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.