Learn More
Waters Corp WATERS CORPORATION Solid Cap, 22 mL Vial, PTFE/Silicone Liner
Catalog No. 50466000 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid cap is designed for 22 mL vials and features a PTFE/Silicone liner. It is used for sample storage and is intended to prevent contamination.
  • Includes cap & septum
  • PTFE silicone lining is resistant to contamination
  • Avoids the need to restart sample preparation procedures due to an unclean surface
  • Designed for proper sample storage
  • Comes in a 100-pack
  • Cap color: White
Catalog No. 50-466-000 Supplier Waters Corp Supplier No. 186004109
This solid cap is designed for 22 mL vials and features a PTFE/Silicone liner. It is used for sample storage and is intended to prevent contamination.
  • Includes cap & septum
  • PTFE silicone lining is resistant to contamination
  • Avoids the need to restart sample preparation procedures due to an unclean surface
  • Designed for proper sample storage
  • Comes in a 100-pack
  • Cap color: White