Learn More
Sigma Aldrich Fine Chemicals Biosciences Solid glass beads borosili
Catalog No. 502905446
Encompass
for use as distillation column packing mixing beads and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. /-10% variation in bead size.
Catalog No. 50-290-5446 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2736274EA
for use as distillation column packing mixing beads and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. /-10% variation in bead size.