Learn More
Sigma Aldrich Fine Chemicals Biosciences Solid-glass beads borosili1EA
Catalog No. 502904002
Encompass
for use as distillation column packing mixing beads and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. /-10% variation in bead size.
Catalog No. 50-290-4002 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1439441EA
for use as distillation column packing mixing beads and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. /-10% variation in bead size.