Learn More
Fisher Science Education™ Digital Dry Bath Solid Heat Block
SCI_ED
Catalog No. S29041
Change view
Click to view available options
No. of Places:
Solid Block
Catalog No. No. of Places
S29041 Solid Block
1 options
Catalog No. S29041 Supplier Fisher Science Education™ Supplier No. BSW01
Add to Cart
Add to Cart
The high-grade aluminum solid heat block is precision machined to provide unsurpassed temperature uniformity.
  • Solid block for slides, petri dishes, or custom matching.
  • High-grade aluminum
  • Dimensions: 7.62 x 9.53 x 5.08cm (3 x 3.75 x 2 in.)
Order Info
Blocks are sold separately from Dry Bath; see S29028 or S29029 for associated Dry Bath.
Certifications
CE
Specifications
For Use With (Equipment) Fisher Digital Dry Baths
Material Aluminum
No. of Places Solid Block
Type Block
For Use With (Application) Heating and incubating samples
Warranty 2 Years
Dimensions (L x W x H) 7.62 x 9.53 x 5.08 cm (3 x 3.75 x 2 in.)