Learn More
GA International Solid Ink Water-Resistant Big Tip Marker Purple 3/pack
Catalog No. 5000561856
Encompass_Preferred
Water-resistant big tip cryo marker. Provided in an assortment of colors (e.g. blue, red, green, orange and yellow) these permanent ink marker can withstand a range of temperatures (-196°C/-321°F to 200°C/392°F). Easy to use, the solid ink markers work on most surfaces, dry in 5-7 minutes and will not fade.
Catalog No. 50-005-61856 Supplier GA International Supplier No. CW1PU3
Water-resistant big tip cryo marker. Provided in an assortment of colors (e.g. blue, red, green, orange and yellow) these permanent ink marker can withstand a range of temperatures (-196°C/-321°F to 200°C/392°F). Easy to use, the solid ink markers work on most surfaces, dry in 5-7 minutes and will not fade.