Learn More
Roboz Surgical SOLID INSTRUMENT TRAY
SDP
Catalog No. NC0243663
Encompass
Solid Instrument Tray; 13.5 X 9.75 X .75 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles
Catalog No. NC0243663 Supplier Roboz Surgical Supplier No. RT1351S
Solid Instrument Tray; 13.5 X 9.75 X .75 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles