Learn More
Trajan Scientific PEEK Tubing, Solid Layer Color Coded PEEK Tubing (Red) - 1/16" OD x .005" ID x 100FT, 6700 PSI, 460 BAR
Catalog No. 501385093
Encompass_Preferred
PEEK Tubing, Solid Layer Color Coded PEEK Tubing (Red) - 1/16" OD x .005" ID x 100FT, 6700 PSI, 460 BAR. PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting, and is not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.
Catalog No. 50-138-5093 Supplier Trajan Scientific Supplier No. VAL.JRT6007100
PEEK Tubing, Solid Layer Color Coded PEEK Tubing (Red) - 1/16" OD x .005" ID x 100FT, 6700 PSI, 460 BAR. PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting, and is not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel. The polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample. Note however that dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.