Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, Side Arm
GREENER_CHOICESDP
Catalog No. 31501582 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-501-582 -
31-501-579 10 mL
31-501-580 25 mL
31-501-581 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-501-582 Supplier Synthware™ Supplier No. P140100M
Add to Cart
Add to Cart
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint and side connection.
Specifications
For Use With (Application) Organic Synthesis
Product Type Vessel with T-bore PTFE stopcock and ground joint
Material Borosilicate Glass