Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, T-bore PTFE Stopcock
GREENER_CHOICESDP
Catalog No. 31501574 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
100 mL
Product Type:
Vessel, Coarse Porosity
Vessel, Medium Porosity
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type
31-501-574 10 mL Vessel, Coarse Porosity
31-501-575 50 mL Vessel, Coarse Porosity
31-501-576 50 mL Vessel, Medium Porosity
31-501-577 100 mL Vessel, Coarse Porosity
31-501-578 100 mL Vessel, Medium Porosity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-501-574 Supplier Synthware™ Supplier No. P130010C
Add to Cart
Add to Cart
Solid Phase Peptide Synthesis Vessel with a side arm and a coarse or medium porosity fritted glass disc.
Specifications
Product Type Vessel, Coarse Porosity
For Use With (Application) Organic Synthesis
Capacity (Metric) 10 mL
Material Borosilicate Glass