Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Solid Phase Peptide Synthesis Vessel, T-bore, Vacuum
GREENER_CHOICESDP

Catalog No. 31501568 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Product Type:
Porosity: Coarse
Porosity: Medium
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type
31-501-568 10 mL Porosity: Coarse
31-501-569 10 mL Porosity: Medium
31-501-570 25 mL Porosity: Coarse
31-501-571 25 mL Porosity: Medium
31-501-572 50 mL Porosity: Coarse
31-501-573 50 mL Porosity: Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-501-568 Supplier Synthware™ Supplier No. P120010C
Add to Cart
Add to Cart

Solid Phase Peptide Synthesis Vessel with fritted glass disc.

Specifications

Product Type Porosity: Coarse
For Use With (Application) Organic Synthesis
Capacity (Metric) 10 mL
Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.