Learn More
Electron Microscopy Sciences Stirring Rod Solid PTFE Rod, 300 mm, diameter
SDP
Catalog No. 5032774
Encompass_Preferred
PTFE solid rod with tapered end, rods are inert and will not scratch. Use up to 280 deg C.
OAL: 300 mm
Catalog No. 50-327-74 Supplier Electron Microscopy Sciences Supplier No. 6620130
PTFE solid rod with tapered end, rods are inert and will not scratch. Use up to 280 deg C.
OAL: 300 mm