Learn More
Grainger Micro 100 Square End Mill, Carbide, Spiral Downcut, 1 ea
Catalog No. 502787865
Encompass_Preferred
This solid carbide router bit features a single-flute, spiral downcut design engineered for efficient finishing operations in plastics and composite materials. Constructed from C2 micro grain carbide, the tool provides durability and consistent performance.
  • Solid carbide construction for enhanced tool longevity
  • Single-flute design with polished flutes to improve chip evacuation
  • Spiral downcut configuration that drives chips downward for clean top surfaces
  • Optimized geometry to support increased material removal rates
  • Square end style with center cutting capability
Catalog No. 50-278-7865 Supplier Grainger Supplier No. 18F075
This solid carbide router bit features a single-flute, spiral downcut design engineered for efficient finishing operations in plastics and composite materials. Constructed from C2 micro grain carbide, the tool provides durability and consistent performance.
  • Solid carbide construction for enhanced tool longevity
  • Single-flute design with polished flutes to improve chip evacuation
  • Spiral downcut configuration that drives chips downward for clean top surfaces
  • Optimized geometry to support increased material removal rates
  • Square end style with center cutting capability