Learn More
Grainger MICRO 100 Solid Carbide Router Bit, Spiral Downcut, 3 mm Cutter, 8 mm LOC, 1 ea.
Catalog No. 502788038
Encompass_Preferred
This single-flute solid carbide router bit features a spiral downcut geometry, engineered specifically for precision routing, trimming, and edge finishing in plastics and composite materials. Its design provides clean, chip-free cuts and prevents edge fraying during aerospace machining and other composite fabrication applications.
  • Single-flute design with a 25° helix angle.
  • Constructed from grade C2 micro-grain solid carbide.
  • Cutter diameter of 3.000 mm with an 8.00 mm length of cut.
  • Overall length of 57.0 mm with a 3.000 mm shank diameter.
  • Center cutting capability.
  • Bright, uncoated finish.
Catalog No. 50-278-8038 Supplier Grainger Supplier No. 18F093
This single-flute solid carbide router bit features a spiral downcut geometry, engineered specifically for precision routing, trimming, and edge finishing in plastics and composite materials. Its design provides clean, chip-free cuts and prevents edge fraying during aerospace machining and other composite fabrication applications.
  • Single-flute design with a 25° helix angle.
  • Constructed from grade C2 micro-grain solid carbide.
  • Cutter diameter of 3.000 mm with an 8.00 mm length of cut.
  • Overall length of 57.0 mm with a 3.000 mm shank diameter.
  • Center cutting capability.
  • Bright, uncoated finish.