Learn More
Supertek™ Solid Rubber Tuning Fork Activator
GREENER_CHOICESCI_ED
Catalog No. S81861 Shop All Supertek Scientific Products
Change view
Click to view available options
Material:
Rubber
Catalog No. Material
S81861 Rubber
1 options
This item is not returnable. View return policy
Catalog No. S81861 Supplier Supertek™ Supplier No. PH36143
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Solid Rubber Tuning Fork Activator is used as a surface to gently strike the tuning fork to activate vibrations. Rubber activator is tapered so the bottom is wider for stability.
  • 1-Quality Tuning Fork Activator
  • Made of Solid Rubber
Specifications
Product Type Tuning Fork Activator
Material Rubber
For Use With (Application) Striking Tuning Forks