Learn More
Grainger Solid Rubber Wheel 10 550 lb.
Catalog No. 502765930
Encompass_Preferred
The MARASTAR Flat Free Solid Rubber Wheel is a 10 inch pneumatic replacement wheel made of solid rubber. It has a load capacity of 1250 lbs and is designed for material handling carts, workbenches, and industrial equipment. The solid rubber construction eliminates flats and punctures. It features precision ball bearings, a sturdy steel hub, and dual grease fittings. The wheel assembly measures 10" x 4" and has a 5/8" inner diameter bearing with a 1/2" thread size. It meets ANSI standards and is made in the USA.
Catalog No. 50-276-5930 Supplier Grainger Supplier No. 16V333
The MARASTAR Flat Free Solid Rubber Wheel is a 10 inch pneumatic replacement wheel made of solid rubber. It has a load capacity of 1250 lbs and is designed for material handling carts, workbenches, and industrial equipment. The solid rubber construction eliminates flats and punctures. It features precision ball bearings, a sturdy steel hub, and dual grease fittings. The wheel assembly measures 10" x 4" and has a 5/8" inner diameter bearing with a 1/2" thread size. It meets ANSI standards and is made in the USA.