Learn More
Grainger Solid Rubber Wheel 4-3/4 280 lb.
Catalog No. 502766515
Encompass_Preferred
The MARASTAR Flat Free Solid Rubber Wheel is a 10 inch pneumatic replacement wheel made of solid rubber. It has a load capacity of 350 lbs and is designed for material handling carts, workbenches, and industrial equipment. The solid rubber construction eliminates flats and the need to maintain air pressure. It has a 5/8 inch precision ball bearing and a 1 inch width profile. The wheel assembly comes with a zinc-plated steel hub and fasteners for easy installation. This heavy-duty wheel provides vibration dampening and quiet operation.
Catalog No. 50-276-6515 Supplier Grainger Supplier No. 16V340
The MARASTAR Flat Free Solid Rubber Wheel is a 10 inch pneumatic replacement wheel made of solid rubber. It has a load capacity of 350 lbs and is designed for material handling carts, workbenches, and industrial equipment. The solid rubber construction eliminates flats and the need to maintain air pressure. It has a 5/8 inch precision ball bearing and a 1 inch width profile. The wheel assembly comes with a zinc-plated steel hub and fasteners for easy installation. This heavy-duty wheel provides vibration dampening and quiet operation.