Learn More
Perkin Elmer US LLC Solid Sample Handling Kit
Catalog No. 502104162
Encompass_Preferred
This kit includes the following tools: Micro Spatula, Qty. 1 Single Quill Brush, Qty. 12 Tweezers - Qty. 1 Wafer-Handling Tweezers, Qty. 1 CHN Sample Capsule Pedestal, Qty. 1
Catalog No. 50-210-4162 Supplier Perkin Elmer US LLC Supplier No. N2410146
This kit includes the following tools: Micro Spatula, Qty. 1 Single Quill Brush, Qty. 12 Tweezers - Qty. 1 Wafer-Handling Tweezers, Qty. 1 CHN Sample Capsule Pedestal, Qty. 1