Learn More
Perkin Elmer US LLC Solid Sample Holder for LAMBDA Spectrometers
Catalog No. 502104154
Encompass_Preferred
PerkinElmer provides optional cell holders and changers that make it possible for you to measure samples from thin films to highly diffuse powders at fixed or variable angles. With these specialty cell holders, you can extend the types of samples you analyze as well as the types of analysis you perform.
Catalog No. 50-210-4154 Supplier Perkin Elmer US LLC Supplier No. B0152471
PerkinElmer provides optional cell holders and changers that make it possible for you to measure samples from thin films to highly diffuse powders at fixed or variable angles. With these specialty cell holders, you can extend the types of samples you analyze as well as the types of analysis you perform.