Learn More
Ace Glass, Inc. #11 Ace-Thred solid PTFE plug, 7855-708 FETFE O-ring front seal
SDP
Catalog No. NC0129028
Encompass_Preferred
#11 Ace-Thred solid PTFE plug with front seal o-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -012 FETFE o-ring.
Catalog No. NC0129028 Supplier Ace Glass, Inc. Supplier No. 584646
#11 Ace-Thred solid PTFE plug with front seal o-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -012 FETFE o-ring.