Learn More
Uline Solid Top Rackable Pallet - 48 x 40 2200 lb Capacity | Blue | 1 EA
Supplier:  Uline H8165
Encompass
Solid smooth top won't rip bags or cases of product, Smooth and sealed to prevent bacteria growth, No crevices for dirt or grime Easy wipe down, Durable injection-molded plastic, 4-way forklift access, USDA and FDA compliant
Catalog No. NC2939529