Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner

Catalog No. 111020923 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
15 mL Vial
20 or 40 mL Vial
22 mL Vial
7 mL Vial
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-102-0923 7 mL Vial
11-101-0572 20 or 40 mL Vial
11-101-1290 15 mL Vial
11-101-1291 22 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-0923 Supplier MilliporeSigma™ Supelco™ Supplier No. 27152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Known for providing the widest range of chemical compatibility and for being the most temperature tolerant of plastic closures

  • The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
  • Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents. 
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) F217/PTFE
Color Green
Material Melamine Resin
No. per Pack 100
Closure Size 15-425
For Use With (Equipment) 7 mL Vial
Product Type Solid Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.