Learn More
MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner
Shop All MilliporeSigma Supelco ProductsDescription
- The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
- Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Specifications
Specifications
| Material (Liner) | F217/PTFE |
| Material | Polypropylene |
| No. per Pack | 100 |
| Quantity | 100 Ea. |
| Closure Size | 10-425 |
| For Use With (Equipment) | 2 mL Large Opening Vial |