Learn More
MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner
Shop All MilliporeSigma Supelco ProductsDescription
- The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
- Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Specifications
Specifications
| Material (Liner) | F217/PTFE |
| Color | Green |
| Material | Melamine Resin |
| No. per Pack | 100 |
| Quantity | 100 Ea. |
| Closure Size | 13-425 |
| For Use With (Equipment) | 4 mL Vial |
| Product Type | Solid Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.