Learn More
BRANSON ULTRASONICS CORPORATION (3P) SOLID TRAY 5.5 GAL
Catalog No. NC3721417
Encompass_Preferred
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 8800 ultrasonic bath, Capacity: 5.5 gal., 20.8L, Depth: 6 in., 15.24cm, Dimensions: 17.5 x 9.5 in. (44.45 x 24.13cm), Includes: Built in handle, Length: 17.5 in., 44.45cm, Stainless Steel
Catalog No. NC3721417 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410178
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 8800 ultrasonic bath, Capacity: 5.5 gal., 20.8L, Depth: 6 in., 15.24cm, Dimensions: 17.5 x 9.5 in. (44.45 x 24.13cm), Includes: Built in handle, Length: 17.5 in., 44.45cm, Stainless Steel