Learn More
Chemglass Life Sciences Solid Waste Container, 5L
SDP
Catalog No. 501311101
Encompass_Preferred
Solid Waste Container, 5L. Container is made of rugged HPDE and has a detachable base and lid with a gasket. It is an ideal container for solid waste in the lab such as used silica gel radioisotopes, contaminated gloves, Kimwipes, filter papers, and other disposables. Overall height with the lid open is 21, footprint is 13 square and has a capacity of 5 Liters. Poly Liner bags are sold separately.
Catalog No. 50-131-1101 Supplier Chemglass Life Sciences Supplier No. CG175970
Solid Waste Container, 5L. Container is made of rugged HPDE and has a detachable base and lid with a gasket. It is an ideal container for solid waste in the lab such as used silica gel radioisotopes, contaminated gloves, Kimwipes, filter papers, and other disposables. Overall height with the lid open is 21, footprint is 13 square and has a capacity of 5 Liters. Poly Liner bags are sold separately.