Learn More
Medchemexpress LLC HY-A0034 10mg Medchemexpress, Solifenacin CAS:242478-37-1 Purity:>98%
SDP
Catalog No. 502082311
Encompass_Preferred
Medchemexpress, HY-A0034 10mg Solifenacin CAS:242478-37-1 Solifenacin (YM905 free base) is a novel muscarinic receptor antagonist with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2311 Supplier Medchemexpress LLC Supplier No. HYA003410MG
Medchemexpress, HY-A0034 10mg Solifenacin CAS:242478-37-1 Solifenacin (YM905 free base) is a novel muscarinic receptor antagonist with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.