Learn More
Selleck Chemical LLC Solifenacin (YM905) S5238-25mg
SDP
Catalog No. 503110333
Encompass_Preferred
Solifenacin (YM905) is a competitive muscarinic receptor antagonist The binding of acetylcholine to these receptors particularly the M3 receptor subtype plays a critical role in the contraction of smooth muscle
Catalog No. 50-311-0333 Supplier Selleck Chemical LLC Supplier No. S523825MG
Solifenacin (YM905) is a competitive muscarinic receptor antagonist The binding of acetylcholine to these receptors particularly the M3 receptor subtype plays a critical role in the contraction of smooth muscle