Learn More
Medchemexpress LLC Solifenacin (free base) | 242478-37-1 | MFCD18910847 | 99.6% | 362.5 g/mol | C23H26N2O2 | 10 MG
SDP
Catalog No. 5000297917
Encompass_Preferred
Solifenacin (Standard) is an analytical reference standard of solifenacin (free base) supplied as a solid powder for research and analytical applications. It is intended for use in assay development, method validation, and quality control in laboratory settings.
  • High purity suitable for analytical use.
  • Supplied as a stable solid powder for easy handling and storage.
  • Accompanied by a certificate of analysis with batch-specific data.
  • Appropriate for assay development and method validation.
  • Matches CAS RN 242478-37-1; molecular weight 362.5 g/mol.
Catalog No. 50-002-97917 Supplier Medchemexpress LLC Supplier No. HYA0034R10MG
Solifenacin (Standard) is an analytical reference standard of solifenacin (free base) supplied as a solid powder for research and analytical applications. It is intended for use in assay development, method validation, and quality control in laboratory settings.
  • High purity suitable for analytical use.
  • Supplied as a stable solid powder for easy handling and storage.
  • Accompanied by a certificate of analysis with batch-specific data.
  • Appropriate for assay development and method validation.
  • Matches CAS RN 242478-37-1; molecular weight 362.5 g/mol.