Learn More
Medchemexpress LLC Solifenacin hydrochloride | 180468-39-7 | 99.7% | 398.93 | C23H27ClN2O2 | 10 MG
SDP
Catalog No. 5000296968
Encompass_Preferred
Solifenacin hydrochloride is a muscarinic receptor antagonist with pKis of 7.6, 6.9, and 8.0 for M1, M2, and M3 receptors, respectively. It reduces bladder responses and shows slight inhibitory effects on salivary and cardiac responses, and is used in research on muscarinic receptor pharmacology and urinary bladder function.
  • High purity suitable for research use (assay ~99.7%).
  • Solid, light yellow to yellow physical appearance.
  • Soluble in DMSO and water; ultrasonic assistance recommended for dissolution.
  • Stable when stored sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Available as small solid quantities and concentrated DMSO solutions for in vitro and in vivo studies.
Catalog No. 50-002-96968 Supplier Medchemexpress LLC Supplier No. HYI023010MG
Solifenacin hydrochloride is a muscarinic receptor antagonist with pKis of 7.6, 6.9, and 8.0 for M1, M2, and M3 receptors, respectively. It reduces bladder responses and shows slight inhibitory effects on salivary and cardiac responses, and is used in research on muscarinic receptor pharmacology and urinary bladder function.
  • High purity suitable for research use (assay ~99.7%).
  • Solid, light yellow to yellow physical appearance.
  • Soluble in DMSO and water; ultrasonic assistance recommended for dissolution.
  • Stable when stored sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Available as small solid quantities and concentrated DMSO solutions for in vitro and in vivo studies.