Learn More
Apexbio Technology LLC Solifenacin succinate, 1g. Cas: 242478-38-2 MFCD: MFCD09952287. Muscarinic receptor antagonist
SDP
Supplier:  Apexbio Technology LLC B16141000
Encompass_Preferred
Solifenacin succinate is a quinuclidine and tetrahydroisoquinoline derivative and selective M3 muscarinic antagonist. It is used as a urological agent in the treatment of overactive bladder[1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5275