Learn More
TARGETMOL CHEMICALS INC Solifenacin succinate 500MG
SDP
Catalog No. 502880355
Encompass_Preferred
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Solifenacin succinate (YM905) is a quinuclidine and tetrahydroisoquinoline derivative and selective M3 MUSCARINIC ANTAGONIST. It is used as a UROLOGIC AGENT in the treatment of URINARY INCONTINENCE. Purity 99.73%
Catalog No. 50-288-0355 Supplier TARGETMOL CHEMICALS INC Supplier No. T0111500MG
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Solifenacin succinate (YM905) is a quinuclidine and tetrahydroisoquinoline derivative and selective M3 MUSCARINIC ANTAGONIST. It is used as a UROLOGIC AGENT in the treatment of URINARY INCONTINENCE. Purity 99.73%