Learn More
Cayman Chemical Solithromycin, Purity: ≥98%, CAS: 760981-83-7, 10 mg
SDP
Catalog No. 502985910
Encompass_Preferred
A macrolide antibiotic; active against Gram-positive and Gram-negative bacteria, including C. trachomatis and C. pneumoniae (MICs = 0.125-0.5 and 0.25-1 μg/ml, respectively); inhibits growth of multidrug resistant P. falciparum in vitro (IC50s = 0.5 nM for both Dd2 and 7G8 strains); eradicates P. berghei infection in mice when administered in a four-dose series at 100 mg/kg
Catalog No. 50-298-5910 Supplier Cayman Chemical Supplier No. 2168110 MG
A macrolide antibiotic; active against Gram-positive and Gram-negative bacteria, including C. trachomatis and C. pneumoniae (MICs = 0.125-0.5 and 0.25-1 μg/ml, respectively); inhibits growth of multidrug resistant P. falciparum in vitro (IC50s = 0.5 nM for both Dd2 and 7G8 strains); eradicates P. berghei infection in mice when administered in a four-dose series at 100 mg/kg