Learn More
TARGETMOL CHEMICALS INC SOLITHROMYCIN 25MG
SDP
Catalog No. 502580883
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Solithromycin (OP-1068) is an orally bioavailable antimicrobial agent that inhibits cell viability protein synthesis and growth rate of Streptococcus pneumoniae Staphylococcus aureus Staphylococcus aureus and Haemophilus influenzae with IC50 values of 7.5 ng/mL 40 ng/mL and 125 ng/mL respectively.Solithromycin inhibits protein biosynthesis by binding to the large 50S subunit of the ribosome. purity: 99%
Catalog No. 50-258-0883 Supplier TARGETMOL CHEMICALS INC Supplier No. T233125MG
Also available in 2 mg 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Solithromycin (OP-1068) is an orally bioavailable antimicrobial agent that inhibits cell viability protein synthesis and growth rate of Streptococcus pneumoniae Staphylococcus aureus Staphylococcus aureus and Haemophilus influenzae with IC50 values of 7.5 ng/mL 40 ng/mL and 125 ng/mL respectively.Solithromycin inhibits protein biosynthesis by binding to the large 50S subunit of the ribosome. purity: 99%