Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solketal, 97%

Catalog No. AC153655000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153655000 500 g Glass bottle
AC153651000 100 g Glass bottle
AC153650025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC153655000 Supplier Thermo Scientific Chemicals Supplier No. 153655000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-79-8 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-79-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00063238
InChI Key RNVYQYLELCKWAN-UHFFFAOYSA-N
Synonym solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
PubChem CID 7528
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Melting Point -50°C
Color Colorless
Density 1.0600g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 500 g
Beilstein 19, V,2, 362
Specific Gravity 1.06
Merck Index 15, 5259
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether
Viscosity 11 mPa.s (20°C)
Formula Weight 132.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Solketal, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 202-888-7
RTECSNumber JI0400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.