Learn More
Cayman Chemical Solndn 10mg
SDP
Catalog No. 502912416
Encompass_Preferred
Source Active recombinant C-terminal human IgG1 Fc-tagged B7-H4 expressed in HEK293 cells Amino acids 29-258 MW 52 3 kDa
Catalog No. 50-291-2416 Supplier Cayman Chemical Supplier No. 36433 10MG
Source Active recombinant C-terminal human IgG1 Fc-tagged B7-H4 expressed in HEK293 cells Amino acids 29-258 MW 52 3 kDa