Learn More
Enterprise Technology Solutions Solo Cup Company Hot Cups, Paper, Poly Lined Inside, 16oz, 50/PK, MI
SDP
Catalog No. 5000790031
Encompass_Preferred
Bistro Paper Hot Cups are great for serving hot beverages like coffee, tea or cocoa. The cup interior is lined with polyethylene that acts as a moisture barrier to keep liquid inside the cup and prevent leaking. Smooth rolled rim gives rigidity to the cup and ensures a leak-resistant lid fit. One lid (sold separately) fits five cup sizes (10 oz. squat to 24 oz.) for operational efficiency.
Catalog No. 50-007-90031 Supplier Enterprise Technology Solutions Supplier No. SCC316SI0041
Bistro Paper Hot Cups are great for serving hot beverages like coffee, tea or cocoa. The cup interior is lined with polyethylene that acts as a moisture barrier to keep liquid inside the cup and prevent leaking. Smooth rolled rim gives rigidity to the cup and ensures a leak-resistant lid fit. One lid (sold separately) fits five cup sizes (10 oz. squat to 24 oz.) for operational efficiency.