Learn More
Enterprise Technology Solutions Solo Cup Company Portion Paper Cup, 3.5oz, 100/PK, White
SDP
Catalog No. 5000790664
Encompass_Preferred
Paper cups are treated, pleated and lightweight. They are ideal for dispensing medications and serving outpatients. Capacity for each is 3.5 oz. Paper cups are made with at least 80 percent renewable resources. They meet ASTM D6868 standard for compostability and are compostable in a commercial compost facility.
Catalog No. 50-007-90664 Supplier Enterprise Technology Solutions Supplier No. SCC4502050
Paper cups are treated, pleated and lightweight. They are ideal for dispensing medications and serving outpatients. Capacity for each is 3.5 oz. Paper cups are made with at least 80 percent renewable resources. They meet ASTM D6868 standard for compostability and are compostable in a commercial compost facility.