Learn More
Pyramex™ Solo Safety Glasses
Catalog No. 19817360 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Clear
Gray
Lens Tint:
Clear
Gray
Size:
Jumbo
Regular
No. per Pack:
1 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Size No. per Pack
19-817-360 Gray Gray Regular 1 Ea.
19-151-423 Clear Clear Regular 1 Ea.
19-151-430 Clear Clear Jumbo 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-817-360 Supplier Pyramex™ Supplier No. S520S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight with vented temples for added comfort.
  • Clear lens ideal for general purposes and indoor applications that require impact protection
  • 100% polycarbonate scratch-resistant lenses provide 99% protection from harmful UV-A and UV-B rays
  • Jumbo size fits over prescription lenses
Compliance
Exceed ANSI™ Z87.1-2003 high velocity impact standards.
Warranty and Services
90 days
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Exceed ANSI Z87.1-2003 High Velocity Impact standards.
Lens Tint Gray
Frame Color Gray
No. per Pack 1 Ea.
Size Regular
Temple Style Vented
Lens Material Polycarbonate