Learn More
Sigma Aldrich Fine Chemicals Biosciences SOLu Trypsin Rapid Digesti
Catalog No. 502904369
Encompass
SOLu-Trypsin Rapid Digestion Kit (MSKT0002) consists of 400 ug of SOLUTIONon-stable trypsin (EMS0004) suitable for protein sequencing and 30 mL of Rapid Digestion Buffer (EMS0009) allowing for trypsin digestion in one hour at 60C. Trypsin should be used at a 110 enzymesubstrate ratio in Rapid Digestion Buffer which can be substituted into existing user workflows.
Catalog No. 50-290-4369 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSKT00021KT
SOLu-Trypsin Rapid Digestion Kit (MSKT0002) consists of 400 ug of SOLUTIONon-stable trypsin (EMS0004) suitable for protein sequencing and 30 mL of Rapid Digestion Buffer (EMS0009) allowing for trypsin digestion in one hour at 60C. Trypsin should be used at a 110 enzymesubstrate ratio in Rapid Digestion Buffer which can be substituted into existing user workflows.