Learn More
Sigma Aldrich Fine Chemicals Biosciences SOLu-Trypsin recombinant e
Catalog No. 502904272
Encompass
Trypsin is a pancreatic serine endoprotease. SOLu-Trypsin is made from recombinant trypsin porcine sequence. It does not possess chymotryptic activity.
Catalog No. 50-290-4272 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. EMS00044X100UG
Trypsin is a pancreatic serine endoprotease. SOLu-Trypsin is made from recombinant trypsin porcine sequence. It does not possess chymotryptic activity.