Learn More
BioVendor, LLC SIRCOL SOLUBLE COLLAGEN ASSAY BOVINE 110 ASSAYS
SDP
Catalog No. 501725954
Encompass
TypeColorimetric assayApplicationsCell culture and/or animal studiesShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15–25°C. The glass vial containing collagen standard should be stored at +4°C once opened. Calibration Range2.5 – 12.5 μg using 250μl of Alkali Reagent to recover the collagen bound dye10 – 50 μg using 1000μl of Alkali Reagent to recover the collagen bound dyeLimit of Detection1.0 μg
Catalog No. 50-172-5954 Supplier BioVendor, LLC Supplier No. S1000
TypeColorimetric assayApplicationsCell culture and/or animal studiesShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationAll components are stable for one year, (from Invoice Date), when stored at 15–25°C. The glass vial containing collagen standard should be stored at +4°C once opened. Calibration Range2.5 – 12.5 μg using 250μl of Alkali Reagent to recover the collagen bound dye10 – 50 μg using 1000μl of Alkali Reagent to recover the collagen bound dyeLimit of Detection1.0 μg