Learn More
Cell Biolabs Inc Soluble Collagen Assay Kit, 96 assays
Supplier:  Cell Biolabs Inc MET5016
Encompass_Preferred
Cell Biolabs’ Soluble Collagen Assay Kit provides a convenient colorimetric method for the detection of soluble collagen from cell or tissue samples. First, the unknown samples or collagen standards are added to a 96 well plate and dried down overnight. Then, a Sirius Red reagent is added to stain the [Gly-x-y] triple helix structure of collagen. Finally, the stained collagen is washed with an Acidic Reagent, eluted from the plate with a Basic Reagent, transferred to a new 96 well plate and measured by a plate spectrophotometer. The amount of collagen in the unknown samples is determined by comparing with a predetermined collagen standard curve. The provided reagents are sufficient for the evaluation of 96 assays including standards and unknown samples.
Catalog No. 50-109-8837