Learn More
Cayman Chemical Soluble EpoxID Hydrolase Cel
SDP
Catalog No. 502892279
Encompass_Preferred
Mammalian sEH is a member of the α/β-hydrolase fold family of enzymes that catalyze the hydrolysis of exogenous and endogenous epoxides to vicinal diols. Endogenous substrates for sEH include EETs which exhibit vasodilatory and anti-inflammatory activity. Cayman’s sEH Cell-Based Assay provides a convenient 96-well plate, fluorescence-based method for detecting epoxide hydrolase activity in whole cells. The assay utilizes Epoxy Fluor 7, a sensitive fluorescent substrate for sEH that can be used to monitor the activity of both human and murine enzymes.
Catalog No. 50-289-2279 Supplier Cayman Chemical Supplier No. 600090480 WELL
Mammalian sEH is a member of the α/β-hydrolase fold family of enzymes that catalyze the hydrolysis of exogenous and endogenous epoxides to vicinal diols. Endogenous substrates for sEH include EETs which exhibit vasodilatory and anti-inflammatory activity. Cayman’s sEH Cell-Based Assay provides a convenient 96-well plate, fluorescence-based method for detecting epoxide hydrolase activity in whole cells. The assay utilizes Epoxy Fluor 7, a sensitive fluorescent substrate for sEH that can be used to monitor the activity of both human and murine enzymes.