Learn More
Cayman Chemical Soluble Epoxide Hydrolase hu
SDP
Catalog No. 502890688
Encompass_Preferred
Source: Active recombinant human N-terminal His-tagged sEH expressed in insect cells • Amino acids: 2-555 (full length) • MW: 64 kDa
Catalog No. 50-289-0688 Supplier Cayman Chemical Supplier No. 1001166950 UG
Source: Active recombinant human N-terminal His-tagged sEH expressed in insect cells • Amino acids: 2-555 (full length) • MW: 64 kDa