Learn More
Cayman Chemical Soluble Epoxide Hydrolase mo
SDP
Catalog No. 502890178
Encompass_Preferred
Source: Active recombinant mouse N-terminal His-tagged sEH expressed in insect cells • Amino acids: 2-554 • MW: 64 kDa
Catalog No. 50-289-0178 Supplier Cayman Chemical Supplier No. 1001167050 UG
Source: Active recombinant mouse N-terminal His-tagged sEH expressed in insect cells • Amino acids: 2-554 • MW: 64 kDa