Learn More
Cayman Chemical Soluble Epoxide Hydrolase Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886524
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to the internal region of human sEH • Host: Rabbit • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) sEH • Applications: WB • MW: ~64 kDa
Catalog No. 50-288-6524 Supplier Cayman Chemical Supplier No. 100101461 EA
Immunogen: Synthetic peptide corresponding to the internal region of human sEH • Host: Rabbit • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) sEH • Applications: WB • MW: ~64 kDa