Learn More
Cusabio Technology LLC soluble fibrin monomer complex
Catalog No. 5000049223
Encompass_Preferred
This kit detects SFMC (soluble fibrin monomer complex) in human serum plasma and urine samples using a sandwich ELISA method With a detection range of 6 25 g/mL to 400 g/mL and a sensitivity of 1 56 g/mL it enables precise quantitative analysis The assay requires a sample volume of 50-100 L and typically completes within 1-5 hours Results are measured at an optical density of 450 nm providing reliable detection of SFMC levels for research or diagnostic applications Its broad sample compatibility and robust performance make it suitable for studying fibrin formation and degradation processes in various clinical or experimental settings
Catalog No. 50-000-49223 Supplier Cusabio Technology LLC Supplier No. CSBE09097H
This kit detects SFMC (soluble fibrin monomer complex) in human serum plasma and urine samples using a sandwich ELISA method With a detection range of 6 25 g/mL to 400 g/mL and a sensitivity of 1 56 g/mL it enables precise quantitative analysis The assay requires a sample volume of 50-100 L and typically completes within 1-5 hours Results are measured at an optical density of 450 nm providing reliable detection of SFMC levels for research or diagnostic applications Its broad sample compatibility and robust performance make it suitable for studying fibrin formation and degradation processes in various clinical or experimental settings