Learn More
Electron Microscopy Sciences Solujet Low Foaming Phosphate Free Liquid Detergent 1 GL
SDP
Supplier:  Electron Microscopy Sciences 6053601
Encompass_Preferred
Low foaming phosphate free liquid cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. This detergent passes the inhibitory residue test. Dilute 1:200 and it has a pH 11.5.
Catalog No. 50-366-290